Abnova
Manufacturer Code:MAB16124
Catalog # 89-999-008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9H5 |
Immunogen | Recombinant protein corresponding to rat Exoc3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex component 3; Exocyst complex component Sec6; rSec6; Sec6; SEC6-like 1; Sec6l1
Gene Aliases: Exoc3; rSec6; Sec6; Sec6l1
UniProt ID: (Rat) Q62825
Entrez Gene ID: (Rat) 252881
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.