Novus Biologicals
Manufacturer Code:NBP271554V
Catalog # NBP271554V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B9 |
Immunogen | Full length recombinant protein of human Eukaryotic translation initiation factor 1 (NP_005792) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: A121; A121Protein translation factor SUI1 homolog eIF1 EIF-1 EIF1A eukaryotic translation initiation factor 1 ISO1 Sui1iso1 SUI1sui1iso1; eIF1; Eukaryotic translation initiation factor 1; Protein translation factor SUI1 homolog; Sui1iso1
Gene Aliases: A121; EIF-1; EIF1; EIF1A; ISO1; SUI1
UniProt ID: (Human) P41567
Entrez Gene ID: (Human) 10209
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.