Novus Biologicals
Manufacturer Code:NBP266826
Catalog # NBP266826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM92-32 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1; Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1 ets protein ETS-1 EWSR2 FLJ10768 p54 protein C-ets-1 v-ets avian erythroblastosis virus E2 oncogene homolog 1 v-ets avian erythroblastosis virus E26 oncogene homolog 1 v-ets erythroblastosis virus E26 oncogene homolog 1 (avian); p54; Protein C-ets-1; v-ets avian erythroblastosis virus E2 oncogene homolog 1; v-ets avian erythroblastosis virus E26 oncogene homolog 1
Gene Aliases: c-ets-1; ETS-1; ETS1; EWSR2; p54
UniProt ID: (Human) P14921
Entrez Gene ID: (Human) 2113
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.