Invitrogen
Manufacturer Code:PA568806
Catalog # PIPA568806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse Estrogen Receptor beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ER beta; ER-beta; estrogen receptor 2 beta; Estrogen receptor beta; Estrogen receptor beta ER-beta Nuclear receptor subfamily 3 group A member 2 ESR2 ESTRB NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta
Gene Aliases: ER-beta; ER[b]; Erb2; ERbeta; Esr2; Estrb; Nr3a2
UniProt ID: (Mouse) O08537, (Rat) Q62986
Entrez Gene ID: (Mouse) 13983, (Rat) 25149
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.