Abnova
Manufacturer Code:PAB16900
Catalog # 89-118-713
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to full length mouse Esrp1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against full-length recombinant Esrp1.
For Research Use Only
Protein Aliases: 2210008M09Rik; A630065D16; BC031468; Epithelial splicing regulatory protein 1; Rbm35a; RNA binding motif protein 35A; RNA-binding motif protein 35A; RNA-binding protein 35A; RP23-322G9.1
Gene Aliases: 2210008M09Rik; A630065D16; BC031468; Esrp1; Rbm35a
UniProt ID: (Mouse) Q3US41
Entrez Gene ID: (Mouse) 207920
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.