R&D Systems
Manufacturer Code:FAB3072P
Catalog # FAB3072P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 38421 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Ala25-Pro250 Accession # P19235 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EPO-R; EpoR EPO-R erythropoietin receptor MGC138358; Erythropoietin receptor
Gene Aliases: EPO-R; EPOR
UniProt ID: (Human) P19235
Entrez Gene ID: (Human) 2057
Molecular Function:
cytokine receptor
receptor
type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.