Abnova
Manufacturer Code:PAB15779
Catalog # 89-117-843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 192 amino acids of mouse Erc1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Erc1.
For Research Use Only
Protein Aliases: 4930404L01Rik; 5033405M01Rik; 9630025C19Rik; B430107L16Rik; CAST2; CAZ-associated structural protein 2; ELKS; ELKS/Rab6-interacting/CAST family member 1; ERC-1; mKIAA1081; Rab6 interacting protein 2; RAB6-interacting protein 2; Rab6ip2; RAB6IP2A; RAB6IP2B
Gene Aliases: 4930404L01Rik; 5033405M01Rik; 9630025C19Rik; B430107L16Rik; CAST2; ELKS; Erc1; Kiaa1081; mKIAA1081; Rab6ip2; RAB6IP2A; RAB6IP2B
UniProt ID: (Mouse) Q99MI1
Entrez Gene ID: (Mouse) 111173
Molecular Function: G-protein modulator enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.