Invitrogen
Manufacturer Code:14968795
Catalog # 14-968-795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HFR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 4ICD; 4ICD ALS19 avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4 avian erythroblastosis oncogene B 4 c-erbB-4 erb-b2 receptor tyrosine kinase 4 ERBB4 ERBB4 intracellular domain ERBB4 transcript variant I12DEL ERBB4 transcript variant I20DEL HER4 human epidermal growth factor receptor 4 Mer4 OTTHUMP00000209432 p180erbB4 Proto-oncogene-like protein c-ErbB-4 receptor tyrosine kinase receptor tyrosine-protein kinase erbB-4 s80HER4 Tyro-2 Tyrosine kinase-type cell surface receptor HER4 v-erb-a erythroblastic leukemia viral oncogene homolog 4 v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) v-erb-b2 avian erythroblastic leukemia viral oncogene 4 v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4; avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4; avian erythroblastosis oncogene B 4; ERBB4 intracellular domain; ERBB4 transcript variant I12DEL; ERBB4 transcript variant I20DEL; human epidermal growth factor receptor 4; p180erbB4; Proto-oncogene-like protein c-ErbB-4; Receptor tyrosine-protein kinase erbB-4; s80HER4; Tyrosine kinase-type cell surface receptor HER4; v-erb-a erythroblastic leukemia viral oncogene homolog 4; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4
Gene Aliases: ALS19; c-erbB-4; ERBB4; HER4; Mer4; p180erbB4
UniProt ID: (Human) Q15303, (Mouse) Q61527
Entrez Gene ID: (Human) 2066, (Mouse) 13869
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.