Novus Biologicals
Manufacturer Code:NBP290781V
Catalog # NB077853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 49 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse ErbB4/Her4 (Q99P91 Met1-Leu652). |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 4ICD; avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4; avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4 EC 2.7.10 EC 2.7.10.1 HER4MGC138404 p180erbB4 Proto-oncogene-like protein c-ErbB-4 receptor tyrosine-protein kinase erbB-4 Tyrosine kinase-type cell surface receptor HER4 v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4 v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian); ERBB4 intracellular domain; ERBB4 transcript variant I12DEL; ERBB4 transcript variant I20DEL; human epidermal growth factor receptor 4; p180erbB4; Proto-oncogene-like protein c-ErbB-4; Receptor tyrosine-protein kinase erbB-4; s80HER4; Tyrosine kinase-type cell surface receptor HER4; v-erb-a erythroblastic leukemia viral oncogene homolog 4; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4
Gene Aliases: ALS19; ERBB4; HER4; p180erbB4
UniProt ID: (Human) Q15303
Entrez Gene ID: (Human) 2066
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.