Novus Biologicals
Manufacturer Code:NBP1049700.025ML
Catalog # NB018277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The antiserum was produced against synthesized phosphopeptide derived from human Her3/ErbB3 around the phosphorylation site of tyrosine 1328 (P-D-Yp-W-H). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-erbB3 EC 2.7.10 EC 2.7.10.1 ErbB-3 erbB3-S HER3c-erbB-3 LCCS2 lethal congenital contracture syndrome 2 MDA-BF-1 MGC88033 p180-ErbB3 p45-sErbB3 p85-sErbB3 Proto-oncogene-like protein c-ErbB-3 receptor tyrosine-protein kinase erbB-3 Tyrosine kinase-type cell surface receptor HER3 v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian); human epidermal growth factor receptor 3; Proto-oncogene-like protein c-ErbB-3; Receptor tyrosine-protein kinase erbB-3; Tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3
Gene Aliases: c-erbB-3; c-erbB3; ErbB-3; ERBB3; erbB3-S; HER3; LCCS2; MDA-BF-1; p180-ErbB3; p45-sErbB3; p85-sErbB3
UniProt ID: (Human) P21860
Entrez Gene ID: (Human) 2065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.