Invitrogen
Manufacturer Code:MA513008
Catalog # PIMA513008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H3.105.5 (Ab105) |
Immunogen | Extracellular domain of recombit human c-erbB-3/HER-3 oncoprotein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: avian erythroblastosis oncogene B 3 receptor; c-erbB-3; Glial growth factor receptor; human epidermal growth factor receptor 3; Proto-oncogene-like protein c-ErbB-3; proto-oncogene-like protein c-ErbB-3 receptor tyrosine-protein kinase erbB-3 tyrosine kinase-type cell surface receptor HER3 v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; receptor tyrosine kinase; Receptor tyrosine-protein kinase erbB-3; Tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3
Gene Aliases: c-erbB-3; c-erbB3; C76256; ErbB-3; ERBB3; erbB3-S; Erbb3r; HER3; LCCS2; MDA-BF-1; p180-ErbB3; p45-sErbB3; p85-sErbB3
UniProt ID: (Human) P21860, (Mouse) Q61526
Entrez Gene ID: (Human) 2065, (Mouse) 13867
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.