Invitrogen
Manufacturer Code:MA513105
Catalog # PIMA513105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | e2-4001 |
Immunogen | Cytoplasmic domain of recombit human c-erbB-2/HER-2 oncoprotein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); c-erb B2/neu protein; C-erbB-2; CD340; epidermal growth factor receptor-related protein; herstatin; human epidermal growth factor receptor 2; Metastatic lymph node gene 19 protein; MLN 19; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; p185erbB2; p185neu; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Receptor tyrosine-protein kinase erbB-2; Receptor tyrosine-protein kinase erbB-2 Metastatic lymph node gene 19 protein MLN 19 Proto-oncogene Neu Proto-oncogene c-ErbB-2 Tyrosine kinase-type cell surface receptor HER2 p185erbB2 CD340 MLN19 NEU NGL c-erb B2/neu protein herstatin metas; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog
Gene Aliases: c-erbB2; c-neu; CD340; Erbb-2; ERBB2; HER-2; HER-2/neu; HER2; Kiaa3023; mKIAA3023; MLN 19; MLN19; NEU; NGL; TKR1
UniProt ID: (Human) P04626, (Mouse) P70424
Entrez Gene ID: (Human) 2064, (Mouse) 13866, (Rat) 24337
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.