Novus Biologicals
Manufacturer Code:NBP271701B
Catalog # NBP271701B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G3 |
Immunogen | Full length recombinant protein of human Epsin-2 (NP_055779) produced in HEK293T cells. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EPS-15-interacting protein 2; Eps15 binding protein; Eps15 binding protein EPS-15-interacting protein 2 epsin 2 epsin-2 KIAA1065EHB21; Epsin-2
Gene Aliases: EHB21; EPN2; KIAA1065
UniProt ID: (Human) O95208
Entrez Gene ID: (Human) 22905
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.