Invitrogen
Manufacturer Code:PA512090
Catalog # PIPA512090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 6-36 amino acids from the N-terminal region of human Epsin2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EPS-15-interacting protein 2; Eps15 binding protein; Epsin 2 EPS-15 interacting protein 2 EPN2 KIAA1065; Epsin-2; Ibp2; Intersectin-EH-binding protein 2
Gene Aliases: 9530051D10Rik; AA536924; EHB21; EPN2; Ibp2; KIAA1065
UniProt ID: (Human) O95208, (Mouse) Q8CHU3
Entrez Gene ID: (Human) 22905, (Mouse) 13855
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.