Invitrogen
Manufacturer Code:MA530526
Catalog # PIMA530526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 19 |
Immunogen | Recombit Mouse Ephrin B3/EFNB3 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: EFL6 EPLG8 LERK8 EPH-related receptor transmembrane ligand ELK-L3 Ephrin B3 eph-related receptor tyrosine kinase ligand 8; Ephrin-B3
Gene Aliases: EFL-6; Efnb3; ELF-3; Elk-L3; Epl8; LERK-8; NLERK-2
UniProt ID: (Mouse) O35393
Entrez Gene ID: (Mouse) 13643
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.