Novus Biologicals
Manufacturer Code:NBP290798AF350
Catalog # NB078102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 19 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse Ephrin-B3 (NP_031937.1 Met1-Ala227). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 8; EPH-related receptor tyrosine kinase ligand 8 Ephrin B3 ephrin-B3 EPLG8EPH-related receptor transmembrane ligand ELK-L3 LERK-8 LERK8EFL6; Ephrin-B3; LERK-8
Gene Aliases: EFL6; EFNB3; EPLG8; LERK8
UniProt ID: (Human) Q15768
Entrez Gene ID: (Human) 1949
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.