R&D Systems
Manufacturer Code:BAF395
Catalog # BAF395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human Ephrin-B3 Leu28-Ser224 Accession # Q15768 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EFL-6 Efnb3 Elk-L3 EPH-related receptor tyrosine kinase ligand 8 Ephrin B3 ephrin-B3 EphrinB3 Epl8 EPLG8EPH-related receptor transmembrane ligand ELK-L3 LERK-8 LERK8EFL6 NLERK-2; EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 8; Ephrin-B3; LERK-8
Gene Aliases: EFL6; EFNB3; EPLG8; LERK8
UniProt ID: (Human) Q15768
Entrez Gene ID: (Human) 1949
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.