Invitrogen
Manufacturer Code:PA538067
Catalog # PIPA538067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic non-phosphopeptide derived from human Ephrin B around the phosphorylation site of Tyr330 (N-I-Y-Y-K) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CEK5 ligand CEK5 receptor ligand CEK5-L CEL5-L EFL-3 EFNB1 ELK ligand ELK-L EPH-related receptor tyrosine kinase ligand 2 EPL2 EPLG2 Ephrin-B1 precursor LERK-2 LERK2 STRA1 STRA1 protein kinase ephrin-B1; CEK5 receptor ligand; CEK5-L; EFL-3; ELK ligand; ELK-L; EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 2; EPH-related receptor tyrosine kinase ligand 8; ephrin ligand B3; Ephrin-B1; Ephrin-B1 C-terminal fragment; Ephrin-B1 CTF; Ephrin-B1 ICD; Ephrin-B1 intracellular domain; Ephrin-B3; LERK-2; LERK-8; Stimulated by retinoic acid gene 1 protein
Gene Aliases: Cek5-L; CFND; CFNS; EFB1; EFL-3; EFL-6; EFL3; EFL6; EFNB1; EFNB3; ELF-3; Elk-L; ELK-L3; Epl2; Epl8; EPLG2; EPLG8; LERK-2; LERK-8; LERK2; LERK8; NLERK-2; Stra1
UniProt ID: (Human) P98172, (Human) Q15768, (Mouse) P52795, (Mouse) O35393
Entrez Gene ID: (Human) 1947, (Human) 1949, (Rat) 25186, (Mouse) 13641, (Mouse) 13643, (Rat) 360546
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.