R&D Systems
Manufacturer Code:MAB3692100
Catalog # MAB3692100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 80937 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Ephrin-A4 Leu26-Gly171 Accession # P52798 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
Protein Aliases: EFL4 EFL-4 EFNA4 EPH-related receptor tyrosine kinase ligand 4 ephrin-A4 EphrinA4 EPLG4MGC125826 LERK-4 LERK4FLJ57652 ligand of eph-related kinase 4; EPH-related receptor tyrosine kinase ligand 4; Ephrin-A4; LERK-4; ligand of eph-related kinase 4
Gene Aliases: EFL4; EFNA4; EPLG4; LERK4
UniProt ID: (Human) P52798
Entrez Gene ID: (Human) 1945
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.