Novus Biologicals
Manufacturer Code:NBP212279H
Catalog # NBP212279H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0262-7N12 |
Immunogen | Human Ephrin A3 |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EFL-2; EFL2 EFL-2 EHK1-L EPH-related receptor tyrosine kinase ligand 3 ephrin-A3 EPLG3EHK1 ligand LERK3LERK-3 ligand of eph-related kinase 3; EHK1 ligand; EHK1-L; EPH-related receptor tyrosine kinase ligand 3; Ephrin-A3; LERK-3; ligand of eph-related kinase 3
Gene Aliases: EFL2; EFNA3; Ehk1-L; EPLG3; LERK3
UniProt ID: (Human) P52797
Entrez Gene ID: (Human) 1944
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.