Novus Biologicals
Manufacturer Code:NBP298317C
Catalog # NB053351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Ephrin-A3 (NP_034238.1 Met1-Ser205). |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EFL-2; EFL2 EFL-2 EHK1-L EPH-related receptor tyrosine kinase ligand 3 ephrin-A3 EPLG3EHK1 ligand LERK3LERK-3 ligand of eph-related kinase 3; EHK1 ligand; EHK1-L; EPH-related receptor tyrosine kinase ligand 3; Ephrin-A3; LERK-3; ligand of eph-related kinase 3
Gene Aliases: EFL2; EFNA3; Ehk1-L; EPLG3; LERK3
UniProt ID: (Human) P52797
Entrez Gene ID: (Human) 1944
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.