Proteintech
Manufacturer Code:124801AP150UL
Catalog # 50-172-8011
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Ephrin A3 Fusion Protein Ag3188 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EFL 2 EFL2 EFNA3 Ehk1 L EHK1 ligand ephrin A3 EPLG3 LERK 3 LERK3; EFL-2; EHK1 ligand; EHK1-L; EPH-related receptor tyrosine kinase ligand 3; Ephrin-A3; LERK-3; ligand of eph-related kinase 3
Gene Aliases: AW494418; EFL-2; EFL2; EFNA3; Ehk1-L; Epl3; EPLG3; LERK-3; LERK3
UniProt ID: (Human) P52797, (Mouse) O08545
Entrez Gene ID: (Human) 1944, (Rat) 170901, (Mouse) 13638
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.