Invitrogen
Manufacturer Code:PA577394
Catalog # PIPA577394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KITHSPQAHDNPQEK corresponding to amino acid residues 150-164 of human Ephrin-A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EPH-related receptor tyrosine kinase ligand 1; Ephrin-A1; Ephrin-A1, secreted form; Immediate early response protein B61; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; Tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4
Gene Aliases: AI325262; B61; ECKLG; EFL1; EFNA1; Epgl1; Epl1; EPLG1; LERK-1; LERK1; TNFAIP4
UniProt ID: (Human) P20827, (Mouse) P52793, (Rat) P97553
Entrez Gene ID: (Human) 1942, (Mouse) 13636, (Rat) 94268
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.