R&D Systems
Manufacturer Code:AF467
Catalog # AF467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Val27-Lys548 Accession # P54763 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CAPB Cek5 Drt DRTEphB2 EC 2.7.10 EC 2.7.10.1 EK5 elk-related tyrosine kinase EPH receptor B2 eph tyrosine kinase 3 EPH-like kinase 5 ephrin type-B receptor 2 EPHT3MGC87492 EPTH3 Erk ERKHek5 HEK5 Nuk PCBC protein-tyrosine kinase HEK5 Qek2 Renal carcinoma antigen NY-REN-47 Sek3 Tyro5 Tyrosine-protein kinase receptor EPH-3 Tyrosine-protein kinase TYRO5; Developmentally-regulated Eph-related tyrosine kinase; EK5; ELK-related tyrosine kinase; EPH tyrosine kinase 3; EPH-like kinase 5; EphB2/CTF1; EphB2/CTF2; Ephrin type-B receptor 2; protein-tyrosine kinase HEK5; Renal carcinoma antigen NY-REN-47; Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase TYRO5
Gene Aliases: CAPB; DRT; EK5; EPHB2; EPHT3; EPTH3; ERK; HEK5; PCBC; TYRO5
UniProt ID: (Human) P29323
Entrez Gene ID: (Human) 2048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.