Invitrogen
Manufacturer Code:PA514577
Catalog # PIPA514577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 896-928 amino acids from the C-terminal region of human EphA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EK4; EPH receptor A3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; Ephrin type-A receptor 3 Tyrosine-protein kinase receptor ETK1 HEK HEK4; HEK; Human embryo kinase; human embryo kinase 1; testicular tissue protein Li 64; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase TYRO4
Gene Aliases: EK4; EPHA3; ETK; ETK1; HEK; HEK4; I79_002835; TYRO4
UniProt ID: (Human) P29320
Entrez Gene ID: (Human) 2042, (Chinese hamster) 100768107
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.