Novus Biologicals
Manufacturer Code:NBP298383AF700
Catalog # NB082350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant EphA3 (EDK98238.1 Gly569-Val984). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 EK4 EPH receptor A3 EphA3 EPH-like kinase 4 ephrin type-A receptor 3 ETK hEK4 HEKETK1eph-like tyrosine kinase 1 human embryo kinase 1 TYRO4 TYRO4 protein tyrosine kinase Tyrosine-protein kinase receptor ETK1 Tyrosine-protein kinase TYRO4; EK4; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; HEK; Human embryo kinase; human embryo kinase 1; testicular tissue protein Li 64; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase TYRO4
Gene Aliases: EK4; EPHA3; ETK; ETK1; HEK; HEK4; TYRO4
UniProt ID: (Human) P29320
Entrez Gene ID: (Human) 2042
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.