R&D Systems
Manufacturer Code:MAB639SP
Catalog # MAB639SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 233720 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse EphA2 Ala22-Ala535 (predicted) Accession # AAA82113 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ARCC2 EC 2.7.10 EC 2.7.10.1 Eck ECKepithelial cell receptor protein tyrosine kinase EPH receptor A2 EphA2 ephrin type-A receptor 2 Epithelial cell kinase Myk2 Sek2 soluble EPHA2 variant 1 Tyrosine-protein kinase receptor ECK; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; soluble EPHA2 variant 1; Tyrosine-protein kinase receptor ECK
Gene Aliases: ARCC2; CTPA; CTPP1; CTRCT6; ECK; EPHA2
UniProt ID: (Human) P29317
Entrez Gene ID: (Human) 1969
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.