Invitrogen
Manufacturer Code:PA514575
Catalog # PIPA514575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 919-950 amino acids from the C-terminal region of human EphA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EPH receptor A2; Ephrin type-A recepor 2 Tyrosine-protein kinase receptor ECK Epithelial cell kinase; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; soluble EPHA2 variant 1; Tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; Tyrosine-protein kinase receptor SEK-2
Gene Aliases: ARCC2; AW545284; CTPA; CTPP1; CTRCT6; ECK; EPHA2; I79_019726; Myk2; Sek-2; Sek2
UniProt ID: (Human) P29317, (Mouse) Q03145
Entrez Gene ID: (Human) 1969, (Chinese hamster) 100752851, (Mouse) 13836
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.