Novus Biologicals
Manufacturer Code:NB1001763
Catalog # NB1001763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5D2 |
Immunogen | Ni-NTA purified truncated recombinant EphA1 expressed in E. Coli strain BL21 (DE3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 EPH receptor A1 eph tyrosine kinase 1 EphA1 EPHephrin type-A receptor 1 EPHT EPHT1erythropoietin-producing hepatoma amplified sequence MGC163163 oncogene EPH soluble EPHA1 variant 1 soluble EPHA1 variant 2 Tyrosine-protein kinase receptor EPH; EPH tyrosine kinase; EPH tyrosine kinase 1; Ephrin type-A receptor 1; erythropoietin-producing hepatoma amplified sequence; Erythropoietin-producing hepatoma receptor; hEpha1; oncogene EPH; soluble EPHA1 variant 1; soluble EPHA1 variant 2; Tyrosine-protein kinase receptor EPH
Gene Aliases: EPH; EPHA1; EPHT; EPHT1
UniProt ID: (Human) P21709
Entrez Gene ID: (Human) 2041
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.