Invitrogen
Manufacturer Code:MA531120
Catalog # PIMA531120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 43 |
Immunogen | Recombit Mouse EPHA1/EPH receptor A1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Embryonic stem cell kinase; EPH EPHT EPHT1 MGC163163; Ephrin type-A receptor 1; mEpha1; Tyrosine-protein kinase receptor ESK
Gene Aliases: 5730453L17Rik; AL033318; Eph; Epha1; Esk
UniProt ID: (Mouse) Q60750
Entrez Gene ID: (Mouse) 13835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.