Invitrogen
Manufacturer Code:PA547435
Catalog # PIPA547435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse EphA1 Glu27-Glu548 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Embryonic stem cell kinase; EPH EPHT EPHT1 eph tyrosine kinase 1 ephrin type-A receptor 1 erythropoietin-producing hepatoma amplified sequence erythropoietin-producing hepatoma receptor hEpha1 oncogene EPH soluble EPHA1 variant 1 soluble EPHA1 variant 2 tyrosine-protein kinase receptor EPH; Ephrin type-A receptor 1; mEpha1; Tyrosine-protein kinase receptor ESK
Gene Aliases: 5730453L17Rik; AL033318; Eph; Epha1; Esk
UniProt ID: (Mouse) Q60750
Entrez Gene ID: (Mouse) 13835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.