Abnova
Manufacturer Code:PAB12333
Catalog # 89-115-726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to rat Epas1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 200&muL of deionized water and concentration will be 500&mu/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Epas1.
For Research Use Only
Protein Aliases: Endothelial PAS domain-containing protein 1; EPAS-1; HIF-2 alpha; HIF-2-alpha; HIF2 alpha; HIF2-alpha; Hif2a; HLF; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia-inducible factor 2 alpha; Hypoxia-inducible factor 2-alpha
Gene Aliases: Epas1; Hif2a; HLF
UniProt ID: (Rat) Q9JHS1
Entrez Gene ID: (Rat) 29452
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.