Novus Biologicals
Manufacturer Code:NBP30335320UL
Catalog # NB093988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human Epac2 (NP_001093867.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP-GEFII; CAMP-GEFII cAMP-regulated guanine nucleotide exchange factor II CGEF2EPAC 2 EPAC2 Exchange factor directly activated by cAMP 2 Exchange protein directly activated by cAMP 2 Nbla00496 putative protein product of Nbla00496 Rap guanine nucleotide exchange factor (GEF) 4 rap guanine nucleotide exchange factor 4 RAP guanine-nucleotide-exchange factor (GEF) 4; cAMP-regulated guanine nucleotide exchange factor II; EPAC 2; Exchange factor directly activated by cAMP 2; Exchange protein directly activated by cAMP 2; putative protein product of Nbla00496; Rap guanine nucleotide exchange factor (GEF) 4; Rap guanine nucleotide exchange factor 4; RAP guanine-nucleotide-exchange factor (GEF) 4
Gene Aliases: CAMP-GEFII; CGEF2; EPAC; EPAC 2; EPAC2; Nbla00496; RAPGEF4
UniProt ID: (Human) Q8WZA2
Entrez Gene ID: (Human) 11069
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.