Novus Biologicals
Manufacturer Code:NBP227361SS
Catalog # NBP227361SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 130-180 of human EpCAM was used as the immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-1A 323/A3 ACSTD1 antigen identified by monoclonal AUA1 CD326 antigen Cell surface glycoprotein Trop-1 chromosome 4 surface marker (35kD glycoprotein) DIAR5 EGP EGP-2 EGP314 EGP40 EpCAM epithelial cell adhesion molecule Epithelial cell surface antigen Epithelial glycoprotein Epithelial glycoprotein 314 ESA GA733-2EGP34 hEGP314 HNPCC8 KS 1/4 antigen KS1/4 KSAHEA125 M1S2 M4S1Ly74 Major gastrointestinal tumor-associated protein GA733-2 MIC18MH99 MOC31 TACST-1 TACSTD1 TROP1CD326 Tumor-associated calcium signal transducer 1CO-17A; Adenocarcinoma-associated antigen; CD326; Cell surface glycoprotein Trop-1; EGP; EGP314; Ep-CAM; Epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; human epithelial glycoprotein-2; KS 1/4 antigen; KSA; Major gastrointestinal tumor-associated protein GA733-2; membrane component, chromosome 4, surface marker (35kD glycoprotein); Tumor-associated calcium signal transducer 1
Gene Aliases: DIAR5; EGP-2; EGP314; EGP40; EPCAM; ESA; GA733-2; HNPCC8; KS1/4; KSA; M1S2; M4S1; MIC18; MK-1; TACSTD1; TROP1
UniProt ID: (Human) P16422
Entrez Gene ID: (Human) 4072
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.