Invitrogen
Manufacturer Code:PA578930
Catalog # PIPA578930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of mouse Eotaxin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 11; Eosinophil chemotactic protein; Eotaxin; Eotaxin C-C motif chemokine 11 Eosinophil chemotactic protein Small-inducible cytokine A11 Ccl11 Scya11; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A11; Small-inducible cytokine A11
Gene Aliases: Ccl11; eotaxin; Scya11
UniProt ID: (Rat) P97545, (Mouse) P48298
Entrez Gene ID: (Rat) 29397, (Mouse) 20292
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.