Invitrogen
Manufacturer Code:PA546946
Catalog # PIPA546946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse CCL11/Eotaxin His24-Pro97 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 11; C-C motif chemokine 11 Eosinophil chemotactic protein Small-inducible cytokine A11 CCL11 SCYA11; Eosinophil chemotactic protein; Eotaxin; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; Small-inducible cytokine A11
Gene Aliases: Ccl11; eotaxin; Scya11
UniProt ID: (Mouse) P48298
Entrez Gene ID: (Mouse) 20292
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.