Invitrogen
Manufacturer Code:PA5114955
Catalog # PIPA5114955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CCL11 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 11; C-C motif chemokine 11 C-C motif chemokine ligand 11 Ccl11 chemokine (C-C motif) ligand 11 eosinophil chemotactic protein Eotaxin Eotaxin 1 Eotaxin1 eotaxin-1 MGC22554 Scya11 small chemokine (C-C motif) ligand 11 small inducible cytokine A11 small inducible cytokine subfamily A (Cys-Cys) member 11 (eotaxin) small inducible cytokine subfamily A11 small-inducible cytokine A11; chemokine (C-C motif) ligand 11; Eosinophil chemotactic protein; Eotaxin; eotaxin-1; small chemokine (C-C motif) ligand 11; small inducible cytokine A11; small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin); small inducible cytokine subfamily A11; Small-inducible cytokine A11
Gene Aliases: CCL11; eotaxin; SCYA11
UniProt ID: (Human) P51671, (Rat) P97545, (Mouse) P48298
Entrez Gene ID: (Human) 6356, (Rat) 29397, (Mouse) 20292
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.