Invitrogen
Manufacturer Code:PA578938
Catalog # PIPA578938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Eotaxin 3 recombit protein (Position: T24-L94). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 26; C-C motif chemokine 26 CC chemokine IMAC Eotaxin-3 Macrophage inflammatory protein 4-alpha MIP-4-alpha Small-inducible cytokine A26 Thymic stroma chemokine-1 TSC-1 CCL26 SCYA26 UNQ216/PRO242; CC chemokine IMAC; chemokine (C-C motif) ligand 26; chemokine N1; Eotaxin-3; Macrophage inflammatory protein 4-alpha; MIP-4-alpha; small inducible cytokine A26; small inducible cytokine subfamily A (Cys-Cys), member 26; Small-inducible cytokine A26; Thymic stroma chemokine-1; TSC-1
Gene Aliases: CCL26; IMAC; MIP-4a; MIP-4alpha; SCYA26; TSC-1; UNQ216/PRO242
UniProt ID: (Human) Q9Y258
Entrez Gene ID: (Human) 10344
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.