Invitrogen
Manufacturer Code:MA523851
Catalog # PIMA523851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 106521 |
Immunogen | E. coli-derived recombit mouse CCL24/Eotaxin-2/MPIF-2 Ile24-Val119 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 24; CC chemokine CCL24; Ckb-6 MPIF-2 MPIF2 SCYA24; Eosinophil chemotactic protein 2; Eotaxin-2; small inducible cytokine A24; Small-inducible cytokine A24
Gene Aliases: Ccl24; CKb-6; MPIF-2; Scya24
UniProt ID: (Mouse) Q9JKC0
Entrez Gene ID: (Mouse) 56221
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.