Invitrogen
Manufacturer Code:PA571861
Catalog # PIPA571861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 109-137 amino acids from the Central region of human C10orf63 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Enkurin; Enkurin ENKUR C10orf63
Gene Aliases: 4933434I06Rik; AU016229; C10orf63; CFAP106; ENKUR
UniProt ID: (Human) Q8TC29, (Mouse) Q6SP97
Entrez Gene ID: (Human) 219670, (Mouse) 71233
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.