Invitrogen
Manufacturer Code:PA533499
Catalog # PIPA533499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from N-terminal extracellular domain of human EDNRB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDNRB ABCDS Endothelin B receptor Etb receptor ET-B Et-b receptor Et-b-r ET-RB ETBR ETRB Etb-svr WS4A Endothelin receptor type B ET-BR ETB HSCR HSCR2; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; ET-B
Gene Aliases: ABCDS; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETRB; HSCR; HSCR2; WS4A
UniProt ID: (Human) P24530
Entrez Gene ID: (Human) 1910
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.