Invitrogen
Manufacturer Code:PA527173
Catalog # PIPA527173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 8 and 111 of Endothelin B Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ednra Etb ET-B ET-BR endothelin B receptor endothelin receptor non-selective type endothelin-B receptor; endothelin B receptor; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; endothelin-B receptor; ET-B; ET-BR; piebald
Gene Aliases: ABCDS; Ednra; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETR-b; ETRB; HSCR; HSCR2; Sox10m1; WS4A
UniProt ID: (Human) P24530, (Rat) P21451, (Mouse) P48302
Entrez Gene ID: (Human) 1910, (Rat) 50672, (Mouse) 13618
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.