Invitrogen
Manufacturer Code:PA124566
Catalog # PIPA124566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 72/72&percnt. Rat: 72/72&percnt. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU022549 ET-B ET>B< ETR-b ETb Sox10m1 s; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; ET-B
Gene Aliases: ABCDS; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETRB; HSCR; HSCR2; WS4A
UniProt ID: (Human) P24530
Entrez Gene ID: (Human) 1910
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.