Invitrogen
Manufacturer Code:PA530589
Catalog # PIPA530589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 363 and 427 of Human Endothelin A Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endothelin A receptor; endothelin receptor subtype A; Endothelin receptor type A; Endothelin-1 receptor; endothelin-1-specific receptor; ET-A; ET-A ETA ETA-R ETAR ETRA hET-AR G protein-coupled receptor endothelin receptor subtype A endothelin-1 receptor endothelin-1-specific receptor; G protein-coupled receptor; G-protein coupled receptor 10
Gene Aliases: EDNRA; ET-A; ET-AR; ETA; ETA-R; ETAR; ETRA; Gpcr10; hET-AR; MFDA
UniProt ID: (Human) Q8TBH2, (Mouse) Q61614
Entrez Gene ID: (Human) 1909, (Mouse) 13617
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.