Novus Biologicals
Manufacturer Code:NBP30350820UL
Catalog # NB095502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 17-187 of human Endothelin-3 (NP_996917.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endothelin 3 endothelin-3 ET3 ET-3 HSCR4 MGC15067 MGC61498 PPET3 preproendothelin-3 truncated endothelin 3 WS4B; Endothelin-3; ET-3; PPET3; Preproendothelin-3
Gene Aliases: EDN3; ET-3; ET3; HSCR4; PPET3; WS4B
UniProt ID: (Human) P14138
Entrez Gene ID: (Human) 1908
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.