Novus Biologicals
Manufacturer Code:NB300526AF532
Catalog # NB30526A532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TR.ET.48.5 |
Immunogen | Pure endothelin-1 conjugated to KLH. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Big endothelin-1; endothelin 1 endothelin-1 ET1 HDLCQ7 PPET1 Preproendothelin-1; Endothelin-1; ET-1; PPET1; Preproendothelin-1
Gene Aliases: ARCND3; EDN1; ET1; HDLCQ7; PPET1; QME
UniProt ID: (Human) P05305
Entrez Gene ID: (Human) 1906
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.