R&D Systems
Manufacturer Code:MAB3440SP
Catalog # MAB3440SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 3G10 |
Immunogen | Human Endothelin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ARCND3 EDN1 endothelin 1 endothelin-1 ET1 HDLCQ7 PPET1 Preproendothelin-1 QME; Big endothelin-1; Endothelin-1; ET-1; PPET1; Preproendothelin-1
Gene Aliases: ARCND3; EDN1; ET1; HDLCQ7; PPET1; QME
UniProt ID: (Human) P05305
Entrez Gene ID: (Human) 1906
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.