Invitrogen
Manufacturer Code:PA3067
Catalog # PIPA3067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse Endothelin-1 CS-Abu-SSLMDKE-Abu-VYF-Abu-HLDIIW coupled to KLH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Big endothelin-1; Endothelin-1; ET-1; PPET1; PPET1 endothelin-1 preproendothelin preproendothelin-1; preproendothelin; Preproendothelin-1
Gene Aliases: ARCND3; EDN1; ET-1; ET1; HDLCQ7; PPET1; preproET; QME
UniProt ID: (Human) P05305, (Rat) P22388, (Mouse) P22387
Entrez Gene ID: (Human) 1906, (Rat) 24323, (Mouse) 13614
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.