Novus Biologicals
Manufacturer Code:NBP121710UV
Catalog # NBP121710UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0079-5J44 |
Immunogen | Mouse recombinant Endocan |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: endocan endothelial cell-specific molecule 1 ESM-1; Endothelial cell-specific molecule 1; ESM-1
Gene Aliases: endocan; ESM1
UniProt ID: (Human) Q9NQ30
Entrez Gene ID: (Human) 11082
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.