R&D Systems
Manufacturer Code:MAB1999
Catalog # MAB1999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 220008 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Endocan/ESM-1 Trp20-Arg184 Accession # Q9QYY7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: endocan endothelial cell-specific molecule 1 ESM1 ESM-1 IGFBP-rp6; Endothelial cell-specific molecule 1; ESM-1
Gene Aliases: endocan; ESM1
UniProt ID: (Human) Q9NQ30
Entrez Gene ID: (Human) 11082
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.